Learn More
Eppendorf™ RackScan
Catalog No. 05413994 Shop All Eppendorf North America Products
Change view
Click to view available options
Type:
Scanner (Bottom Reading)
Scanner (Side Reading)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
05-413-994 Scanner (Side Reading)
05-413-993 Scanner (Bottom Reading)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 05-413-994 Supplier Eppendorf™ Supplier No. 6002000014
Add to Cart
Add to Cart
RackScan reads complete storage racks with up to 96 different barcoded vials. The sample IDs are transferred to downstream databases.
  • Camera based instrument resulting in rapid imaging and decoding
  • Comfortable and easy plug-in accessory
  • Small footprint, ideal for benchtop (and integration)
  • ECC 200 error correction function enables safe code reading with up to 30 % damaged codes
  • Support material of vials available via Eppendorf Homepage for documentation purposes
  • Plug & Transfer with Eppendorf™ eLABInventory and eLABJournal software for comfortable data transfer
  • RackScan s can be attached to RackScan b as accessory to read barcodes on side labels
  • Different formats usable for high flexibility
TRUSTED_SUSTAINABILITY
Specifications
Description RackScan s
Data Transfer Method PC/USB
For Use With (Application) Scanning more than one vial in parallel and transferring the sample data to your sample management software
For Use With (Equipment) SBS racks and cryostorage boxes
Type Scanner (Side Reading)
Connection Type USB