Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Ractopamine Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA175375
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA175375 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA175375 Supplier Invitrogen™ Supplier No. PA175375
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

Working Concentration: 2.5 μg/mL. Sensitivity: 10 ng/mL Ractopamine produces 72% inhibition in a competitive ELISA employing Ractopamine polyclonal antibody.

Ractopamine is a phenolethanolamine b-adrenoceptor agonist that is used for the improvement of weight gain, carcass leanness and feed efficiency in pigs. It has been known for years that ractopamine enhances growth in meat animals by repartitioning nutrients. This means that the nutrients that the animal consumes in its feed are shifted away from being stored as fat and are instead used in lean tissue production. This shift occurs as the animal reaches the top of its growth curve and lean tissue production drops off and fat deposition increases. Fat levels also appear to decrease, due to an increase in lypolysis (fat breakdown). Using the ractopamine monoclonal antibody, it is possible to detect ractopamine and other agonists in urine, muscle and liver both rapidly and with accuracy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ractopamine
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Alias C18H23NO3
Host Species Sheep
Immunogen Ractopamine(10)-BSA.
Purification Method Purified
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.