Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rad1 Antibody, DyLight 650, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297717C
Description
Rad1 Polyclonal specifically detects Rad1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
Rad1 | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence | |
cell cycle checkpoint protein Hrad1, checkpoint control protein HRAD1, DNA repair exonuclease rad1 homolog, DNA repair exonuclease REC1, EC 3.1.11.2, exonuclease homolog RAD1, HRAD1, RAD1 (S. pombe) homolog, RAD1 homolog (S. pombe), Rad1-like DNA damage checkpoint protein, REC1cell cycle checkpoint protein RAD1 | |
Produced in rabbits immunized with E. coli-derived Human Rad1 fragment. | |
0.1 mL | |
Cancer, DNA Repair, Signal Transduction | |
5810 | |
Store at 4°C in the dark. | |
IgG |
Immunofluorescence | |
DyLight 650 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction