Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RAD1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552326
Description
Immunogen sequence: GLREAFSELD MTSEVLQITM SPDKPYFRLS TFGNAGSSHL DYPKDSDLME AFHCNQTQVN RYKISLLKPS TKALVLSCKV SIRTDNRGFL SLQYMIRNED GQICFVEYYC CPDEEVPESE S Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 91%.
Rad1 is a component of a heterotrimeric PCNA-like complex that contains Rad9 and Hus1 proteins. This complex is involved in the 9-1-1 cell cycle checkpoing response complex that plays a major role in DNA repair, by associating with Rad17 and several components of the PCNA-loading heteropentamer replication factor C. Mutants has been shown to partially restore radiation resistance and G2-checkpoint activity. It has also been shown to possess exonuclease activity.
Specifications
RAD1 | |
Polyclonal | |
Unconjugated | |
RAD1 | |
cell cycle checkpoint protein Hrad1; cell cycle checkpoint protein RAD1; checkpoint control protein HRAD1; DNA repair exonuclease rad1 homolog; DNA repair exonuclease REC1; exonuclease homolog RAD1; hRAD1; mRAD1; RAD1; RAD1 checkpoint clamp component; RAD1 checkpoint DNA exonuclease; RAD1 homolog; RAD1 homolog (S. pombe); rad1-like DNA damage checkpoint protein; REC1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5810 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O60671 | |
RAD1 | |
Recombinant protein corresponding to Human RAD1. Recombinant protein control fragment (Product #RP-89473). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction