Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rad17 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NBP186561
Description
Rad17 Polyclonal specifically detects Rad17 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Rad17 | |
Polyclonal | |
Western Blot 0.4 ug/ml, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:200-1:500 | |
CCYC, cell cycle checkpoint protein (RAD17), cell cycle checkpoint protein RAD17, FLJ41520, HRAD17, R24L, RAD1 (S. pombe) homolog, RAD1 homolog, RAD17 homolog (S. pombe), Rad17-like protein, RAD17SP, Rad24, RF-C activator 1 homolog, RF-C/activator 1 homolog | |
Rabbit | |
Affinity Purified | |
RUO | |
5884 | |
Human | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
RAD17 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:SKEHGIQVQEWINPVLPDFQKDDFKGMFNTESSFHMFPYQSQIAVFKEFLLRATKYNKLQMLGDDLRTDKKIILVEDLPNQFYRDSHTLHEVLRKYVRIGRCPLIFIISDSLSGDNNQRLLFPKEIQEECSISNISFNPVAP | |
0.1 mL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction