Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rad17, p Ser645 Antibody, Alexa Fluor™ 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100273AF350
Description
Rad17 Polyclonal antibody specifically detects Rad17 in Human, Mouse samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
Rad17 | |
Polyclonal | |
Western Blot, Immunohistochemistry | |
CCYC, cell cycle checkpoint protein (RAD17), cell cycle checkpoint protein RAD17, FLJ41520, HRAD17, R24L, RAD1 (S. pombe) homolog, RAD1 homolog, RAD17 homolog (S. pombe), Rad17-like protein, RAD17SP, Rad24, RF-C activator 1 homolog, RF-C/activator 1 homolog | |
A synthetic peptide, which represented a portion of human Rad17 (LocusLink ID 5884) around serine 645. | |
0.1 mL | |
Cancer, Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, Phospho Specific | |
5884 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunohistochemistry | |
Alexa Fluor 350 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction