Learn More
Invitrogen™ RAD17 Polyclonal Antibody

Description
PA1-4131 detects yeast RAD17. PA1-4131 has been successfully used in Western blot procedures. By Western blot this antibody detects a ~75 kDa protein representing RAD17 in yeast. The PA1- 4131 immunogen is a synthetic peptide corresponding to residues S(10) T K R S L K K K K I R K I(23) of the 75 kDa S. pombe RAD17.
Specifications
Specifications
| Antigen | RAD17 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | Rad17 |
| Gene Accession No. | P48581 |
| Gene Alias | CCYC; cell cycle checkpoint protein (RAD17); cell cycle checkpoint protein RAD17; DNA damage checkpoint control protein RAD17; DNA repair exonuclease RAD17; FLJ41520; hRad17; MmRad24; R24L; RAD1 homolog; RAD17; RAD17 (Ser647); RAD17 checkpoint clamp loader component; RAD17 homolog; RAD17 homolog (S. pombe); Rad17 like protein; Rad17-like protein; Rad17p; RAD17SP; RAD24; RF-C activator 1 homolog; RF-C/activator 1 homolog; YOR368W |
| Gene Symbols | Rad17 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.