Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAD17 Polyclonal Antibody, QED Bioscience™
SDP

Catalog No. 502278153
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-227-8153 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-227-8153 Supplier Leinco Technologies Inc Supplier No. 56229100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

RAD17 Polyclonal antibody specifically detects RAD17 in Human samples. It is validated for Western Blot

Specifications

Antigen RAD17
Applications Western Blot
Classification Polyclonal
Concentration 0.5-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Rad17
Gene Accession No. O75943
Gene Alias CCYC; cell cycle checkpoint protein (RAD17); cell cycle checkpoint protein RAD17; DNA damage checkpoint control protein RAD17; DNA repair exonuclease RAD17; FLJ41520; hRad17; MmRad24; R24L; RAD1 homolog; RAD17; RAD17 (Ser647); RAD17 checkpoint clamp loader component; RAD17 homolog; RAD17 homolog (S. pombe); Rad17 like protein; Rad17-like protein; Rad17p; RAD17SP; RAD24; RF-C activator 1 homolog; RF-C/activator 1 homolog; YOR368W
Host Species Rabbit
Immunogen Peptide corresponding to aa 2-15 of human RAD17.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5884
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.