Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RAD18 Antibody (79B1048.1), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10056523IR
Description
RAD18 Monoclonal specifically detects RAD18 in Human samples. It is validated for Western Blot.Specifications
RAD18 | |
Monoclonal | |
DyLight 755 | |
EC 6.3.2.-, hHR18, hRAD18, postreplication repair protein hRAD18p, Postreplication repair protein RAD18, RAD18 homolog (S. cerevisiae), RAD18, S. cerevisiae, homolog, RING finger protein 73, RNF73E3 ubiquitin-protein ligase RAD18 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
In HL60, a 54 kDa band should be detected. | |
Store at 4C in the dark. | |
IgG1 |
Western Blot | |
79B1048.1 | |
Western Blot | |
RAD18 | |
The antibody was raised against a synthetic peptide corresponding to the amino acids 402-414 of human Rad18. | |
0.1 mL | |
Core ESC Like Genes, Stem Cell Markers, Zinc Finger | |
56852 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction