Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAD18 Mouse anti-Human, Clone:2G8D12, Abnova™

Catalog No. 89333493 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-333-493 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-333-493 Supplier Abnova Corporation Supplier No. MAB17130
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant human RAD18.

The protein encoded by this gene is highly similar to S. cerevisiae DNA damage repair protein Rad18. Yeast Rad18 functions through its interaction with Rad6, which is an ubiquitin-conjugating enzyme required for post-replication repair of damaged DNA. Similar to its yeast counterpart, this protein is able to interact with the human homolog of yeast Rad6 protein through a conserved ring-finger motif. Mutation of this motif results in defective replication of UV-damaged DNA and hypersensitivity to multiple mutagens. [provided by RefSeq]

Specifications

Antigen RAD18
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 2G8D12
Conjugate Unconjugated
Description RAD18 homolog (S. cerevisiae)
Dilution ELISA (1:10000)
Western Blot (1:500-1:2000)
Immunohistochemistry
Immunocytochemistry
Flow Cytometry (1:200-1:400)
The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene RAD18
Gene Alias RNF73
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acid 332-430 of human RAD18 from E. coli.
Quantity 100 μg
Gene ID (Entrez) 56852
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.