Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAD18 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589233
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589233 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589233 Supplier Invitrogen™ Supplier No. PA589233
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: INKHLDSCLS REEKKESLRS SVHKRKPLPK TVYNLLSDRD LKKKLKEHGL SIQGNKQQLI KRHQEFVHMY NAQCDALHPK SAAEIVREIE NIEKTRMRLE ASKLNESVMV FTKDQTEKEI DEIHSKYRKK HKSEFQLLVD QARKGYKKIA GMSQKTVTIT KEDESTEKLS SVCMGQEDNM TSVTNHFSQS KLDSPEELEP DREEDSSSCI DIQEVLSSSE SDSCNSSSSD IIRDLLEEEE AWEASHKNDL QDTEISPRQN RRTRAAESAE IEPRNKRNRN; Positive Samples: K-562, MCF-7, Jurkat, 293T, Mouse liver; Cellular Location: Cytoplasm, Nucleus, centrosome, cytoskeleton, microtubule organizing center.

Human Rad18 (hRad18) is homologue of the S. pombe Rad18 (3,4). In yeast, Rad6 and Rad18 play a crucial role in post-replication repair functions in gap-filling of a daughter strand on replication of damaged DNA (2). Furthermore, Rad6-Rad18 interaction is thought to be critical at an early step during lesion bypass in yeast. In vivo, Rad18 protein forms a tight complex with Rad6 through a conserved ring-finger motif (3,4). It has been suggested that Rad18-Rad6 complex binds to the damaged sites by the single-stranded DNA binding property of Rad 18. This is followed by degradation of the stalled components of DNA replication by ubiquitin-conjugating enzyme activity of Rad6 (1). The human Rad18 maps on chromosome 3p24-25, the region which is often deleted in lung, breast, ovary, and testis cancers. The human RAD18 gene codes for a protein of 495 amino acids with a predicted molecular weight of 56 kDa. The human Rad18 is ubiquitously expressed in various human tissues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAD18
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.48 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Rad18
Gene Accession No. Q9NS91, Q9QXK2
Gene Alias 2810024C04Rik; E3 ubiquitin-protein ligase RAD18; hHR18; hRAD18; mRAD18Sc; postreplication repair protein hRAD18p; postreplication repair protein RAD18; post-replication repair protein RAD18SC; Rad18; RAD18 E3 ubiquitin protein ligase; RAD18 homolog; RAD18 homolog (S. cerevisiae); RAD18, E3 ubiquitin protein ligase; RAD18, S. cerevisiae, homolog; Rad18sc; RING finger protein 73; RING-type E3 ubiquitin transferase RAD18; RNF73
Gene Symbols Rad18
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 216-495 of human Rad18 (NP_0645503).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 362412, 56852, 58186
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.