Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RAD18 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5118513
Description
Antibody detects endogenous levels of total RAD18.
Human Rad18 (hRad18) is homologue of the S. pombe Rad18 (3,4). In yeast, Rad6 and Rad18 play a crucial role in post-replication repair functions in gap-filling of a daughter strand on replication of damaged DNA (2). Furthermore, Rad6-Rad18 interaction is thought to be critical at an early step during lesion bypass in yeast. In vivo, Rad18 protein forms a tight complex with Rad6 through a conserved ring-finger motif (3,4). It has been suggested that Rad18-Rad6 complex binds to the damaged sites by the single-stranded DNA binding property of Rad 18. This is followed by degradation of the stalled components of DNA replication by ubiquitin-conjugating enzyme activity of Rad6 (1). The human Rad18 maps on chromosome 3p24-25, the region which is often deleted in lung, breast, ovary, and testis cancers. The human RAD18 gene codes for a protein of 495 amino acids with a predicted molecular weight of 56 kDa. The human Rad18 is ubiquitously expressed in various human tissues.
Specifications
| RAD18 | |
| Polyclonal | |
| Unconjugated | |
| Rad18 | |
| 2810024C04Rik; E3 ubiquitin-protein ligase RAD18; hHR18; hRAD18; mRAD18Sc; postreplication repair protein hRAD18p; postreplication repair protein RAD18; post-replication repair protein RAD18SC; Rad18; RAD18 E3 ubiquitin protein ligase; RAD18 homolog; RAD18 homolog (S. cerevisiae); RAD18, E3 ubiquitin protein ligase; RAD18, S. cerevisiae, homolog; Rad18sc; RING finger protein 73; RING-type E3 ubiquitin transferase RAD18; RNF73 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 362412, 56852 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9NS91 | |
| Rad18 | |
| A synthesized peptide derived from human RAD18(Accession Q9NS91), corresponding to amino acid residues N306-Q356. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction