Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rad21 Antibody (CM110-2C10) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$206.50 - $501.50
Specifications
Antigen | Rad21 |
---|---|
Clone | CM110-2C10 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:500 (ECL), Simple Western 1:50 |
Applications | Western Blot |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB100386SS
![]() |
Novus Biologicals
NB100386SS |
0.025 mL |
Each for $206.50
|
|
|||||
NB100386
![]() |
Novus Biologicals
NB100386 |
0.1 mL |
Each for $501.50
|
|
|||||
Description
Rad21 Monoclonal specifically detects Rad21 in Human, Mouse samples. It is validated for Western Blot, Simple Western.Specifications
Rad21 | |
1 mg/mL | |
Western Blot | |
Unconjugated | |
Mouse | |
Apoptosis, Cancer, Cell Biology, Cell Cycle and Replication, Chromatin Research, DNA Repair | |
O60216 | |
5885 | |
An N-terminal fusion protein of the C-terminal fragment of Scc1 subunit human cohesin (amino acids 544-631 of hRad21). | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
CM110-2C10 | |
Western Blot 1:500 (ECL), Simple Western 1:50 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse | |
double-strand-break repair protein rad21 homolog, hHR21FLJ25655, HR21, KIAA0078RAD21 (S. pombe) homolog, MCD1, Nuclear matrix protein 1, NXP-1, NXP1HRAD21, protein involved in DNA double-strand break repair, RAD21 homolog (S. pombe), SCC1 homolog, SCC1FLJ40596 | |
RAD21 | |
IgG1 κ | |
Protein G purified | |
NB100-386 is specific for both the nuclear hRad21 protein and the cleaved cytoplasmic hRad21 protein. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title