Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RAD21 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA575308
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
This gene encodes a nuclear protein that regulates transcriptional elongation and pre-mRNA splicing. The encoded protein interacts with the hyperphosphorylated C-terminal domain of RNA polymerase II via multiple FF domains, and with the pre-mRNA splicing factor SF1 via a WW domain. Alternative splicing results in multiple transcripts variants encoding different isoforms.
Specifications
RAD21 | |
Polyclonal | |
Unconjugated | |
Rad21 | |
64-kDa carboxy-terminal product; 64-kDa C-terminal product; 65-kDa carboxy-terminal product; CDLS4; double-strand-break repair protein rad21 homolog; double-strand-break repair protein rad21-like protein; FLJ25655; FLJ40596; h NXP-1; hHR21; HR21; HRAD21; KIAA0078; KIAA0078RAD21; kleisin; MCD1; mHR21; mKIAA0078; nuclear matrix protein 1; NXP1; NXP-1; Pokeweed agglutinin-binding protein 29; protein involved in DNA double-strand break repair; PW29; Rad21; RAD21 cohesin complex component; RAD21 homolog; RAD21 homolog (S. pombe); SCC1; SCC1 homolog; sister chromatid cohesion 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
19357, 314949, 5885 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
O60216, Q61550 | |
Rad21 | |
Synthetic peptide corresponding to amino acids 521-570 of Human Rad21. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction