Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rad21 Recombinant Mouse Monoclonal Antibody, AbFlex™, Active Motif™

Mouse Recombinant Monoclonal Antibody
$108.45 - $482.00
Specifications
Antigen | Rad21 |
---|---|
Concentration | 1 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-3770
![]() ![]() |
Active Motif
91246 |
10 μg |
Each for $108.45
|
|
|||||
50-199-3769
![]() ![]() |
Active Motif
91245 |
100 μg |
Each for $482.00
|
|
|||||
Description
This gene encodes a nuclear protein that regulates transcriptional elongation and pre-mRNA splicing. The encoded protein interacts with the hyperphosphorylated C-terminal domain of RNA polymerase II via multiple FF domains, and with the pre-mRNA splicing factor SF1 via a WW domain. Alternative splicing results in multiple transcripts variants encoding different isoforms.Specifications
Rad21 | |
Immunocytochemistry, Immunofluorescence, Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse | |
O60216, Q61550 | |
19357, 5885 | |
This antibody was raised against recombinant protein corresponding to the N-terminal half of mouse Rad21. | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
Rad21 |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
140mM HEPES, 32mM sodium acetate with 30% glycerol, 70mM NaCl and 0.035% sodium azide; pH 7.5 | |
64-kDa carboxy-terminal product; 64-kDa C-terminal product; 65-kDa carboxy-terminal product; BOS_14006; CDLS4; Double-strand-break repair protein rad21 homolog; FLJ25655; FLJ40596; h NXP-1; hHR21; Hr21; HRAD21; KIAA0078; KIAA0078RAD21; kleisin; MCD1; mHR21; mKIAA0078; Nuclear matrix protein 1; NXP1; NXP-1; Pokeweed agglutinin-binding protein 29; protein involved in DNA double-strand break repair; PW29; RAD21; RAD21 cohesin complex component; RAD21 homolog; RAD21 homolog (S. pombe); SCC1; SCC1 homolog; sister chromatid cohesion 1 | |
Rad21 | |
IgG2a | |
Protein A | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title