Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RAD21 Recombinant Rabbit Monoclonal Antibody (BLR052F)
Rabbit Recombinant Monoclonal Antibody
$173.10 - $436.00
Specifications
Antigen | RAD21 |
---|---|
Clone | BLR052F |
Concentration | 40 μg/mL |
Content And Storage | 4°C |
Applications | Flow Cytometry, Western Blot, Western Blot |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA544297
|
Invitrogen™
MA544297 |
100 μL |
Each for $436.00
|
|
|||||
PIMA544298
|
Invitrogen™
MA544298 |
20 μL |
Each for $173.10
|
|
|||||
Description
RAD21 Recombinant Monoclonal Antibody for Western Blot, Flow
This gene encodes a nuclear protein that regulates transcriptional elongation and pre-mRNA splicing. The encoded protein interacts with the hyperphosphorylated C-terminal domain of RNA polymerase II via multiple FF domains, and with the pre-mRNA splicing factor SF1 via a WW domain. Alternative splicing results in multiple transcripts variants encoding different isoforms.Specifications
RAD21 | |
40 μg/mL | |
Flow Cytometry, Western Blot, Western Blot | |
Unconjugated | |
Rabbit | |
RUO | |
BBS with 0.1% BSA and 0.09% sodium azide; pH 7.8-8.6 | |
O60216, Q61550 | |
19357, 5885 | |
Peptide representing a region between residue 581 and the C-terminus (residue 631) of human Double-strand-break repair protein Rad21 homolog. | |
Antibody |
BLR052F | |
4°C | |
Recombinant Monoclonal | |
Liquid | |
IgG | |
Human, Mouse | |
Rad21 | |
64-kDa carboxy-terminal product; 64-kDa C-terminal product; 65-kDa carboxy-terminal product; CDLS4; double-strand-break repair protein rad21 homolog; double-strand-break repair protein rad21-like protein; FLJ25655; FLJ40596; h NXP-1; hHR21; HR21; HRAD21; KIAA0078; KIAA0078RAD21; kleisin; MCD1; mHR21; mKIAA0078; nuclear matrix protein 1; NXP1; NXP-1; Pokeweed agglutinin-binding protein 29; protein involved in DNA double-strand break repair; PW29; Rad21; RAD21 cohesin complex component; RAD21 homolog; RAD21 homolog (S. pombe); SCC1; SCC1 homolog; sister chromatid cohesion 1 | |
Rad21 | |
Primary | |
purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title