Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rad23 Antibody (1E4D6) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$560.50
Specifications
Antigen | hHR23A |
---|---|
Clone | 1E4D6 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:500-1:2000, ELISA 1:10000, CyTOF-ready |
Applications | Western Blot, ELISA, CyTOF, Immunohistochemistry, Immunocytochemistry |
Description
Rad23 Monoclonal specifically detects Rad23 in Human samples. It is validated for Western Blot, ELISA, CyTOF-ready.Specifications
hHR23A | |
1 mg/mL | |
Western Blot, ELISA, CyTOF, Immunohistochemistry, Immunocytochemistry | |
Unconjugated | |
Mouse | |
Apoptosis, Cancer, DNA Repair, Nucleotide Excision Repair, Tumor Suppressors | |
PBS with 0.05% Sodium Azide | |
5886 | |
Purified recombinant fragment of human RAD23A (AA: 1-363) expressed in E. Coli. | |
Primary | |
40 kDa |
1E4D6 | |
Western Blot 1:500-1:2000, ELISA 1:10000, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Human | |
hHR23A, HR23A | |
RAD23A | |
IgG2b | |
Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title