Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rad23 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987746
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7746 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7746 Supplier Bioss Supplier No. BS7121R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The RAD23 gene of Saccharomyces cerevisiae is required for excision-repair of UV damaged DNA (1). RAD23 resembles the other DNA repair genes, RAD2, RAD6, RAD7, RAD18, and RAD54, all of which also exhibit increased transcription in response to DNA damage and during meiosis (2). RAD23 encodes a nuclear protein containing a ubiquitin-like domain required for biological function (3). RAD23 bears a ubiquitin-like domain at its amino terminus and this ubiquitin-like domain affects protein function in a nonproteolytic manner (3). Rad23 is a highly conserved protein involved in nucleotide excision repair (NER) that associates with the proteasome via its amino-terminus (4). Its carboxy-terminal ubiquitin-associated domain is evolutionarily conserved from yeast to humans (4). In addition to a role in DNA repair events in yeast, several lines of evidence indicate that the Rad23 protein may regulate the activity of the 26 S proteasome (5).

Specifications

Antigen Rad23
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from Saccharomyces cerevisiae Rad23.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.