Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAD23A Mouse anti-Human, Clone:1E4D6, Abnova™

Catalog No. 89333476 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-333-476 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-333-476 Supplier Abnova Corporation Supplier No. MAB17155
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant human RAD23A.

The protein encoded by this gene is one of two human homologs of Saccharomyces cerevisiae Rad23, a protein involved in nucleotide excision repair (NER). This protein was shown to interact with, and elevate the nucleotide excision activity of 3-methyladenine-DNA glycosylase (MPG), which suggested a role in DNA damage recognition in base excision repair. This protein contains an N-terminal ubiquitin-like domain, which was reported to interact with 26S proteasome, as well as with ubiquitin protein ligase E6AP, and thus suggests that this protein may be involved in the ubiquitin mediated proteolytic pathway in cells. [provided by RefSeq]

Specifications

Antigen RAD23A
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1E4D6
Conjugate Unconjugated
Description RAD23 homolog A (S. cerevisiae)
Dilution ELISA (1:10000)
Western Blot (1:500-1:2000)
Immunohistochemistry
Immunocytochemistry
Flow Cytometry
The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene RAD23A
Gene Alias HHR23A, MGC111083
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acid 1-363 of human RAD23A from E. coli.
Quantity 100 μg
Gene ID (Entrez) 5886
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.