Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RAD23A Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA542125
Description
Peptide sequence: VPSSGSSGRE EDAASTLVTG SEYETMLTEI MSMGYERERV VAALRASYNN Sequence homology: Cow: 93%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
The product of this gene is a small iron-sulfur protein that transfers electrons from NADPH through ferredoxin reductase to a terminal cytochrome P450. This particular oxidation/reduction system is found in steroidogenic tissues, and is involved with the synthesis of bile acid and vitamin D. In addition to the expressed gene at this chromosomal locus (11q22), there are pseudogenes located on chromosomes 20 and 21. This gene product has been identified in a number of different tissues but all forms have been shown to be identical and are not tissue specific.
Specifications
| RAD23A | |
| Polyclonal | |
| Unconjugated | |
| RAD23A | |
| 2310040P19Rik; AL024030; HHR23A; HR23A; MGC111083; Mhr23a; rad23; RAD23 homolog A (S. cerevisiae); RAD23 homolog A, nucleotide excision repair protein; RAD23, yeast homolog, A; RAD23A; RAD23a homolog (S. cerevisiae); UV excision repair protein RAD23 homolog A | |
| Rabbit | |
| Protein A | |
| RUO | |
| 5886 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P54725 | |
| RAD23A | |
| Synthetic peptide directed towards the N-terminal of human RAD23A (aa 141-190). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction