Learn More
Invitrogen™ RAD23A Polyclonal Antibody

Description
Immunogen sequence: LSDDVPIRDY RIDEKNFVVV MVTKTKAGQG TSAPPEASPT AAPESSTSFP PAPTSGMSHP PPAAREDKSP SEESAPTTSP ESVSGSVPSS GSSGREEDAA STLVTGSEYE TMLTEIMSMG YER Highest antigen sequence indentity to the following orthologs: Mouse - 86%, Rat - 89%.
Specifications
Specifications
| Antigen | RAD23A |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | RAD23A |
| Gene Accession No. | P54725 |
| Gene Alias | 2310040P19Rik; AL024030; HHR23A; HR23A; MGC111083; Mhr23a; rad23; RAD23 homolog A (S. cerevisiae); RAD23 homolog A, nucleotide excision repair protein; RAD23, yeast homolog, A; RAD23A; RAD23a homolog (S. cerevisiae); UV excision repair protein RAD23 homolog A |
| Gene Symbols | RAD23A |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.