Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAD26L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA532231
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA532231 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA532231 Supplier Invitrogen™ Supplier No. PA532231
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: SK-N-SH, 293T transfected recombinant C9orf102 protein. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a member of the Snf2 family of helicase-like proteins. The encoded protein may play a role in DNA repair and mitochondrial function. Mutations in this gene have been associated with bone marrow failure syndrome 2. Alternatively spliced transcript variants that encode different protein isoforms have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAD26L
Applications Western Blot
Classification Polyclonal
Concentration 0.62 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene Ercc6l2
Gene Accession No. Q5T890
Gene Alias 0610007P08Rik; 1700019D06Rik; 9330134C04Rik; BMFS2; C8H9orf102; C9orf102; DNA excision repair protein ERCC-6-like 2; DNA repair and recombination protein RAD26-like; ERCC excision repair 6 like 2; Ercc6l2; excision repair cross-complementation group 6 like 2; excision repair cross-complementation group 6-like 2; excision repair cross-complementing rodent repair deficiency, complementation group 6 like 2; excision repair cross-complementing rodent repair deficiency, complementation group 6-like 2; LOW QUALITY PROTEIN: DNA excision repair protein ERCC-6-like 2; putative DNA repair and recombination protein RAD26-like; putative DNA repair and recombination protein RAD26-like-like protein; putative repair and recombination helicase RAD26L; RAD26L; RGD1561537; Sr278; stretch responsive protein 278
Gene Symbols Ercc6l2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human RAD26L. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 375748
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.