Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAD3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA532174
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA532174 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA532174 Supplier Invitrogen™ Supplier No. PA532174
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: Yeast lysate. Predicted reactivity: Yeast (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

ATP-dependent DNA helicase involved in excision repair of DNA damaged with UV light, bulky adducts, or cross-linking agents. Necessary for excision of pyrimidine dimers. Also unwinds DNA/RNA duplexes. Plays an essential role in the cell viability. Involved in the maintenance of the fidelity of DNA replication. Acts as component of the general transcription and DNA repair factor IIH (TFIIH) core, which is essential for both basal and activated transcription, and is involved in nucleotide excision repair (NER) of damaged DNA. TFIIH has CTD kinase and DNA-dependent ATPase activity, and is essential for polymerase II transcription in vitro.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAD3
Applications Western Blot
Classification Polyclonal
Concentration 0.9 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene RAD3
Gene Accession No. P06839
Gene Alias DNA repair helicase RAD3; General transcription and DNA repair factor IIH helicase subunit XPD; RAD3; REM1; RNA polymerase II transcription factor B subunit RAD3; TFB subunit RAD3; TFIIH subunit XPD; TFIIH/NER complex ATP-dependent 5'-3' DNA helicase subunit RAD3; YER171W
Gene Symbols RAD3
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the N-terminus region of Saccharomyces cerevisiae S288c RAD3. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 856918
Target Species Yeast
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.