Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAD50 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA532176
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA532176 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA532176 Supplier Invitrogen™ Supplier No. PA532176
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: Yeast. Predicted reactivity: Yeast (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Rad50-13B3 recognizes human Rad50, a 153a protein involved in DNA double strand break repair. Rad50 is associated with Mre11 and p95 (Nibrin) to form a multiprotein complex involved in the double strand break repair process. Upon DNA damage, Rad50 and Mre11 redistribute and are co-localized within the nucleus of damaged cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAD50
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene RAD50
Gene Accession No. P12753
Gene Alias 153 kDa protein; DNA repair protein RAD50; hRad50; hsRAD50; I79_004319; mRad50; Mrell; MRX complex DNA-binding subunit; N0872; NBSLD; Rad50; RAD50 2; RAD50 double strand break repair protein; RAD50 homolog (S. cerevisiae); RAD50 homolog, double strand break repair protein; RAD50 PEN; RAD502; RAD50-2; RAD50-3; Rad50l; YNL250W
Gene Symbols RAD50
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of Saccharomyces cerevisiae S288c RAD50. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 855471
Target Species Yeast
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.