Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAD50 Rabbit anti-Human, Mouse, Unconjugated, Clone: ACCG-18, Abnova™

Catalog No. 89339760 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-339-760 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-339-760 Supplier Abnova Corporation Supplier No. MAB22022
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody.

RAD50 Monoclonal antibody specifically detects RAD50 in Human, Mouse samples. It is validated for WB, IF.

Specifications

Antigen RAD50
Applications Western Blot, Immunofluorescence
Classification Monoclonal
Clone ACCG-18
Conjugate Unconjugated
Description RAD50 homolog (S. cerevisiae)
Gene RAD50
Gene Alias RAD50-2, hRad50
Host Species Rabbit
Immunogen A synthetic peptide corresponding to human RAD50
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 10111
Target Species Human, Mouse
Content And Storage Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.