Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rad51 Antibody (13E4), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 3 publications
Supplier: Novus Biologicals NB100499
Description
Rad51 Monoclonal specifically detects Rad51 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Rad51 | |
Monoclonal | |
1.0mg/mL | |
Western Blot 1:500-1:1000, Immunohistochemistry | |
RAD51 | |
Recombinant human Rad51 protein. | |
Protein G purified | |
RUO | |
5888 | |
Human | |
Purified |
Western Blot, Immunohistochemistry | |
130000 | |
Unconjugated | |
BRCC5, HRAD51, HsRAD51, HsT16930, RAD51 (S. cerevisiae) homolog (E coli RecA homolog), RAD51 homolog (RecA homolog, E. coli) (S. cerevisiae), RAD51 homolog A, RAD51ABRCA1/BRCA2-containing complex, subunit 5, RecA, E. coli, homolog of, RECADNA repair protein RAD51 homolog 1, RecA-like protein, recombination protein A | |
Mouse | |
37 kDa | |
100 μL | |
Primary | |
This is specific for Rad51. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction