Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rad51 Antibody (523713), Alexa Fluor™ 532, R&D Systems™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals MAB99481AF532
Description
Rad51 Monoclonal specifically detects Rad51 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
Rad51 | |
Monoclonal | |
Alexa Fluor 532 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
RAD51 | |
E. coli-derived partial recombinant human Rad51 protein (amino acids 174-339) | |
0.1 mL | |
Cancer, DNA Repair, Homologous Recombination, modENCODE Antibodies, Phospho Specific | |
Human | |
IgG3 |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
523713 | |
Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
BRCC5, HRAD51, HsRAD51, HsT16930, RAD51 (S. cerevisiae) homolog (E coli RecA homolog), RAD51 homolog (RecA homolog, E. coli) (S. cerevisiae), RAD51 homolog A, RAD51ABRCA1/BRCA2-containing complex, subunit 5, RAD51L3, RecA, E. coli, homolog of, RECADNA repair protein RAD51 homolog 1, RecA-like protein, recombination protein A | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction