Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rad51 Antibody (RAD51/2701), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP279895MFV500
Description
Rad51 Monoclonal antibody specifically detects Rad51 in Human samples. It is validated for Flow CytometrySpecifications
Rad51 | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
RAD51/2701 | |
Flow Cytometry | |
BRCC5, HRAD51, HsRAD51, HsT16930, RAD51 (S. cerevisiae) homolog (E coli RecA homolog), RAD51 homolog (RecA homolog, E. coli) (S. cerevisiae), RAD51 homolog A, RAD51ABRCA1/BRCA2-containing complex, subunit 5, RAD51L3, RecA, E. coli, homolog of, RECADNA repair protein RAD51 homolog 1, RecA-like protein, recombination protein A | |
Recombinant fragment of human Rad51 protein (around aa 1-134) (exact sequence is proprietary) (Uniprot: Q06609) | |
0.1 mL | |
Cancer, DNA Repair, Homologous Recombination, modENCODE Antibodies, Phospho Specific | |
5888 | |
Store at 4C in the dark. | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction