Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RECA-1 Monoclonal Antibody (HIS52 (87.6E4))
GREENER_CHOICE

Catalog No. PIMA123842
Change view
Click to view available options
Quantity:
125 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA123842 125 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA123842 Supplier Invitrogen™ Supplier No. MA123842
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody does not react with mouse, guinea pig, rabbit, porcine, goat, ovine, or chicken protein.

RECA-1 is a cell surface antigen which is expressed by all rat endothelial cells. The endothelium is the thin layer of cells that line the interior surface of blood vessels, forming an interface between circulating blood in the lumen and the rest of the vessel wall. Endothelial cells line the entire circulatory system, from the heart to the smallest capillary.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RECA-1
Applications Immunohistochemistry (Frozen), Immunocytochemistry
Classification Monoclonal
Clone HIS52 (87.6E4)
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide
Gene Rad51
Gene Accession No. 0
Gene Alias Reca1
Gene Symbols Rad51
Host Species Mouse
Immunogen Stromal cells from rat lymph node.
Purification Method Protein G
Quantity 125 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 499870
Target Species Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.