Learn More
Invitrogen™ RAD51 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA592858
Description
Positive Samples: U-937, Mouse brain, Mouse ovary, Mouse testis, Rat testis; Cellular Location: Cytoplasm, Mitochondrion matrix, Nucleus, centrosome, cytoskeleton, microtubule organizing center, perinuclear region Immunogen sequence: MAMQMQLEAN ADTSVEEESF GPQPISRLEQ CGINANDVKK LEEAGFHTVE AVAYAPKKEL INIKGISEAK ADKILAEAAK LVPMGFTTAT EFHQRRSEII QITTGSKELD KLLQGGIETG SITEMFGEFR TGKTQICHTL AVTCQLPIDR GGGEGKAMYI DTEGTFRPER LLAVAERYGL SGSDVLDNVA YARAFNTDHQ TQLLYQASAM MVESRYALLI VDSATALYRT DYSGRGELSA RQMHLARFLR MLLRLADEFG VAVVITNQVV AQVDGAAMFA ADPKKPIGGN IIAHASTTRL YLRKGRGETR ICKIYDSPCL PEAEAMFAIN ADGVGDAKD.
RAD51 plays a critical role in homologous strand exchange, a key step in DNA repair through homologous recobination. It binds to single and double-stranded DNA and exhibits DNA-dependent ATPase activity. RAD51 catalyzes the recognition of homology and strand exchange between homologous DNA partners to form a joint molecule between a processed DNA break and the repair template. It binds to a single-stranded DNA in an ATP-dependent manner to form nucleoprotein filaments which are essential for the homology search and strand exchange. Mutations in the gene can results in breast cancer, mirror movements 2 and fanconi anemia.
Specifications
RAD51 | |
Polyclonal | |
Unconjugated | |
Rad51 | |
AV304093; BRCA1/BRCA2-containing complex, subunit 5; BRCC5; DNA repair protein RAD51; DNA repair protein RAD51 homolog 1; DNA repair protein RAD51 homolog A; DNA repair protein rhp51; FANCR; homolog to S.cerevisiae; HRAD51; HsRad51; HsT16930; MGC128559 protein; MRMV2; MUT5; rad51; RAD51 homolog; RAD51 homolog (RecA homolog); RAD51 homolog (RecA homolog, E. coli); RAD51 homolog A; RAD51 recombinase; RAD51 recombinase L homeolog; rad51.1; rad51.L; rad51-1; RAD51A; rad51-a; rad51-b; Reca; RecA family recombinase Rhp51; RecA, E. coli, homolog of; RecA-like protein; recombinase RAD51; recombination protein A; RGD1563603; rhp51; SPAC644.14c; wu:fb38e12; xrad51; XRad51.1; YER095W; zgc:77754 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
19361, 499870, 5888 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
0.81 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q06609, Q08297 | |
Rad51 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-339 of human Rad51 (NP_0028662). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.