Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RAD51AP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA530395
Description
Recommended positive controls: HeLa, HCT116. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
May participate in a common DNA damage response pathway associated with the activation of homologous recombination and double-strand break repair. Functionally cooperates with PALB2 in promoting of D-loop formation by RAD51. Binds to single and double stranded DNA, and is capable of aggregating DNA. Also binds RNA.
Specifications
RAD51AP1 | |
Polyclonal | |
Unconjugated | |
Rad51ap1 | |
2510006L10Rik; PIR51; RAB22; RAD51 associated protein 1; rad51ap1; RAD51-associated protein 1; RAD51-interacting protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10635 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.61 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q96B01 | |
Rad51ap1 | |
Recombinant protein fragment corresponding to a region within amino acids 1 and 137 of Human Rad51AP1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction