Learn More
Invitrogen™ RAD51AP2 Polyclonal Antibody

Description
Immunogen sequence: VQSVSLISKE VNVEENKYVN QNYVTNTNEY ESILPEREIA NSKDFHRKND SALYINHQFE TGLSEGNDEC FQDLAAKYLS TEA Antigen sequence identity: Mouse 60%; Rat 59%.
Specifications
Specifications
| Antigen | RAD51AP2 |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | RAD51AP2 |
| Gene Accession No. | Q09MP3 |
| Gene Symbols | RAD51AP2 |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.