Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAD51B Monoclonal Antibody (1 H3/13)
GREENER_CHOICE

Catalog No. PIMA524771
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524771 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524771 Supplier Invitrogen™ Supplier No. MA524771
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Does not cross-react with Rad51C, Rad51D, Rad51, XRCC2, or XRCC3 in Western analysis. This antibody does not work in mouse.

Five human RAD 51 homologues have been identified: XRCC2, XRCC3, RAD51B, RAD51C, and RAD51D. Each of these homologues interacts with one or more of the others, with all of the proteins involved in one complex or multiple smaller complexes. Rad51B plays a role in meiotic recombination and/or recombinational repair.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAD51B
Applications Western Blot
Classification Monoclonal
Clone 1 H3/13
Concentration 0.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene Rad51b
Gene Accession No. O15315, O35719
Gene Alias AI553500; DNA repair protein RAD51 homolog 2; hREC2; MGC34245; mREC2; R51H2; R51H2REC2hREC2; RAD51 homolog B; RAD51 paralog B; Rad51B; Rad51l1; RAD51-like 1; RAD51-like protein 1; REC2; RecA-like protein; recombination repair protein; RGD1560681
Gene Symbols Rad51b
Host Species Mouse
Immunogen His-tagged human Rad51L1, overexpressed in E. coli.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19363, 5890
Target Species Human, Mouse, Monkey
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.