Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAD51B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565683
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565683 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565683 Supplier Invitrogen™ Supplier No. PA565683
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SIPVILTNQIT THLSGALASQ ADLVSPADDL SLSEGTSGSS CVIAALGNTW SHSVNTRLIL QYLDSERRQI Highest antigen sequence identity to the following orthologs - mouse 25%, rat 32%.

Five human RAD 51 homologues have been identified: XRCC2, XRCC3, RAD51B, RAD51C, and RAD51D. Each of these homologues interacts with one or more of the others, with all of the proteins involved in one complex or multiple smaller complexes. Rad51B plays a role in meiotic recombination and/or recombinational repair.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAD51B
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Rad51b
Gene Accession No. O15315
Gene Alias AI553500; DNA repair protein RAD51 homolog 2; hREC2; MGC34245; mREC2; R51H2; R51H2REC2hREC2; RAD51 homolog B; RAD51 paralog B; Rad51B; Rad51l1; RAD51-like 1; RAD51-like protein 1; REC2; RecA-like protein; recombination repair protein; RGD1560681
Gene Symbols Rad51b
Host Species Rabbit
Immunogen Recombinant Human RAD51B. Recombinant protein control fragment (Product #RP-106355).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5890
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.