Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rad51C Antibody (2H11/6), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB100177F
Description
Rad51C Monoclonal specifically detects Rad51C in Human, Mouse, Primate, Yeast samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Rad51C | |
Monoclonal | |
FITC | |
O43502 | |
RAD51C | |
His-tagged human Rad51C, over-expressed in E. coli. [UniProt# O43502] | |
Protein G purified | |
RUO | |
Primary | |
Does not cross-react with Rad51B, Rad51D, Rad51, XRCC2, or XRCC3 in Western analysis. | |
Store at 4C in the dark. | |
IgG1 κ |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
2H11/6 | |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
BROVCA3, DNA repair protein RAD51 homolog 3, FANCO, MGC104277, R51H3, RAD51 homolog C, RAD51 homolog C (S. cerevisiae), RAD51L2RAD51 (S. cerevisiae) homolog C, RAD51-like protein 2, yeast RAD51 homolog 3 | |
Mouse | |
40 kDa | |
0.1 mL | |
Breast Cancer, Cancer, Chromatin Research, DNA Double Strand Break Repair, DNA Repair, Homologous Recombination | |
5889 | |
Human, Mouse, Primate, Yeast | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction