Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rad51C Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | Rad51C |
---|---|
Applications | Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
Rad51C Polyclonal specifically detects Rad51C in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
Rad51C | |
Polyclonal | |
Rabbit | |
Breast Cancer, Cancer, Chromatin Research, DNA Double Strand Break Repair, DNA Repair, Homologous Recombination | |
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
5889 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:MRGKTFRFEMQRDLVSFPLSPAVRVKLVSAGFQTAEELLEVKPSELSKEVGISKAEALETLQIIRRECLTNKPRYAGTSESHKKCTALELLEQEHTQGFIITFCSALDDILGGGVPLMKTTEICGAPGVG | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
Human | |
BROVCA3, DNA repair protein RAD51 homolog 3, FANCO, MGC104277, R51H3, RAD51 homolog C, RAD51 homolog C (S. cerevisiae), RAD51L2RAD51 (S. cerevisiae) homolog C, RAD51-like protein 2, yeast RAD51 homolog 3 | |
RAD51C | |
IgG | |
Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title