Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rad51C Antibody (Rad51C 2H11/6), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250263AF750
Description
Rad51C Monoclonal specifically detects Rad51C in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
Rad51C | |
Monoclonal | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
RAD51C | |
Rad51C 2H11/6 was raised against His-tagged human Rad51C, overexpressed in e.coli and purified on a talon affinity column under denaturing conditions, followed by gel purification on a SDS-Page | |
0.1 mL | |
Primary | |
Rad51C 2H11/6 detects as little as 10ng of purified protein and also detects endogenouse hamster Rad51C in whole V79 cell extracts by western blotting. | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
Rad51C 2H11/6 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
BROVCA3, DNA repair protein RAD51 homolog 3, FANCO, MGC104277, R51H3, RAD51 homolog C, RAD51 homolog C (S. cerevisiae), RAD51L2RAD51 (S. cerevisiae) homolog C, RAD51-like protein 2, yeast RAD51 homolog 3 | |
Mouse | |
Protein A purified | |
Breast Cancer, Cancer, Chromatin Research, DNA Double Strand Break Repair, DNA Repair, Homologous Recombination | |
5889.0 | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction