Learn More
RAD51C Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302645AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
This gene is a member of the RAD51 family of related genes, which encode strand-transfer proteins thought to be involved in recombinational repair of damaged DNA and in meiotic recombination. This gene product interacts with two other DNA repair proteins, encoded by RAD51B and XRCC3, but not with itself. The protein copurifies with XRCC3 protein in a complex, reflecting their endogenous association and suggesting a cooperative role during recombinational repair. This gene is one of four localized to a region of chromosome 17q23 where amplification occurs frequently in breast tumors. Overexpression of the four genes during amplification has been observed and suggests a possible role in tumor progression. Alternative splicing has been observed for this gene and two variants encoding different isoforms have been identified.Specifications
RAD51C | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
O43502 | |
RAD51C | |
Between 326 and 376 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
RAD51C | |
DNA repair protein RAD51 homolog 3, RAD51 homolog C (S. cerevisiae), RAD51L2, Yeast RAD51 homolog 3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5889 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.