Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RAD51C Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA577078
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
ZFPM1 is a transcription regulator that plays an essential role in erythroid and megakaryocytic cell differentiation. ZFPM1 is an essential cofactor that acts via the formation of a heterodimer with transcription factors of the GATA family GATA1, GATA2 and GATA3. Such heterodimer can both activate or repress transcriptional activity, depending on the cell and promoter context. The heterodimer formed with GATA proteins is essential to activate expression of genes such as NFE2, ITGA2B, alpha- and beta-globin, while it represses expression of KLF1. ZFPM1 may be involved in regulation of some genes in gonads. ZFPM1 may also be involved in cardiac development, in a non-redundant way with ZFPM2/FOG2.
Specifications
RAD51C | |
Polyclonal | |
Unconjugated | |
RAD51C | |
BROVCA3; DNA repair protein RAD51 homolog 3; FANCO; MGC104277; R51H3; RAD51 homolog C; RAD51 paralog C; RAD51C; RAD51L2; RAD51-like protein 2; RGD1563765; yeast RAD51 homolog 3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
114714, 5889 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
O43502, Q924H5 | |
RAD51C | |
Recombinant full length Human RAD51C. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction