Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rad51D Antibody (SR1590), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP322532100UL
Description
Rad51D Monoclonal antibody specifically detects Rad51D in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, ImmunoprecipitationSpecifications
Rad51D | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry, Immunoprecipitation | |
SR1590 | |
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:50 | |
DNA repair protein RAD51 homolog 4, HsTRAD, R51H3Trad, RAD51 homolog D, RAD51DRAD51 (S. cerevisiae)-like 3, RAD51-like 3 (S. cerevisiae), RAD51-like protein 3, recombination repair protein, TRAD | |
A synthesized peptide derived from human Rad51D (Uniprot #: O75771) | |
100 μg | |
Breast Cancer, Cancer, Chromatin Research, DNA Double Strand Break Repair, DNA Repair, Homologous Recombination | |
5892 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction