Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rad51L1 Antibody (OTI7H4), PE/Atto594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273782PEATT594
Description
Rad51L1 Monoclonal specifically detects Rad51L1 in Human, Mouse samples. It is validated for Flow Cytometry.Specifications
Rad51L1 | |
Monoclonal | |
PE/ATTO 594 | |
PBS with 0.05% Sodium Azide | |
RAD51B | |
Recombinant protein fragment corresponding to amino acids 119-346 of human Rad51L1 (NP_598193) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse | |
IgG1 |
Flow Cytometry | |
OTI7H4 | |
Flow Cytometry | |
DNA repair protein RAD51 homolog 2, MGC34245, R51H2REC2hREC2, RAD51 (S. cerevisiae)-like 1, RAD51 homolog B, RAD51B, RAD51-like 1 (S. cerevisiae), RAD51-like protein 1, RecA-like protein, recombination repair protein | |
Mouse | |
Affinity Purified | |
Breast Cancer, Cancer, Chromatin Research, DNA Double Strand Break Repair, DNA Repair, Homologous Recombination | |
5890.0 | |
Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction