Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RAD52 Antibody (SR1844), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32225450UL
Description
RAD52 Monoclonal antibody specifically detects RAD52 in Human samples. It is validated for Western BlotSpecifications
RAD52 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot | |
SR1844 | |
Western Blot 1:500-1:2000 | |
DNA repair protein RAD52 homolog, RAD52 (S. cerevisiae) homolog, RAD52 homolog (S. cerevisiae), recombination protein RAD52, rhabdomyosarcoma antigen MU-RMS-40.23 | |
A synthesized peptide derived from human RAD52 (Uniprot #: P43351) | |
50 μg | |
Breast Cancer, DNA Repair, Homologous Recombination | |
5893 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction