Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rad53/SPK1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981672
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1672 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1672 Supplier Bioss Supplier No. BS11792R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DNA damage results in the arrest of cell cycle progression, allowing the damaged DNA to be repaired prior to replication. Checkpoints exist at several cell cycle phase transitions to maintain this genetic integrity. Rad9, Rad17, Rad24 and Mec3 are involved in activating the G1 and G2 checkpoints (14). Pol2 (also known as Dun2), encoding the catalytic subunit of DNA polymerase epsilon, plays a role in activating the S phase checkpoint (5). The protein kinase Rad53 (also designated Spk1, Mec2 or Sad1) is essential for both G2 and S phase arrest. Activation of Rad53 is regulated by Mec1 (also known as Esr1 and Sad3), a homolog of the human ATM protein (6). Pds1 and Mad2 both regulate checkpoints associated with incomplete spindle replication (7,8). Dun1, another protein kinase, plays a role in transducing the DNA damage signal (9).

Specifications

Antigen Rad53/SPK1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CHEK2
Gene Accession No. O96017
Gene Symbols CHEK2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Rad53/SPK1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11200
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.