Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RAD54B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA529881
Description
Recommended positive controls: 293T, A431, H1299, HeLa, HepG2, Molt-4, Raji. Predicted reactivity: Bovine (83%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene belongs to the DEAD-like helicase superfamily. It shares similarity with Saccharomyces cerevisiae RAD54 and RDH54, both of which are involved in homologous recombination and repair of DNA. This protein binds to double-stranded DNA, and displays ATPase activity in the presence of DNA. This gene is highly expressed in testis and spleen, which suggests active roles in meiotic and mitotic recombination. Homozygous mutations of this gene were observed in primary lymphoma and colon cancer.
Specifications
RAD54B | |
Polyclonal | |
Unconjugated | |
RAD54B | |
DNA repair and recombination protein RAD54B; E130016E03Rik; fibrinogen silencer-binding protein; FSBP; LOW QUALITY PROTEIN: DNA repair and recombination protein RAD54B; RAD54 homolog B; RAD54 homolog B (S. cerevisiae); RAD54, S. cerevisiae, homolog of, B; Rad54b; RDH54; RGD1306507 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
25788 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q9Y620 | |
RAD54B | |
Recombinant fragment corresponding to a region within amino acids 48 and 291 of Human RAD54B. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction