Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAD54B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541272
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541272 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541272 Supplier Thermo Scientific Supplier No. PA541272
Add to Cart
Add to Cart

RAD54B Polyclonal Antibody

RAD54B Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

TRUSTED_SUSTAINABILITY

Specifications

Antigen RAD54B
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene RAD54B
Gene Accession No. Q9Y620
Gene Alias DNA repair and recombination protein RAD54B; E130016E03Rik; fibrinogen silencer-binding protein; FSBP; LOW QUALITY PROTEIN: DNA repair and recombination protein RAD54B; RAD54 homolog B; RAD54 homolog B (S. cerevisiae); RAD54, S. cerevisiae, homolog of, B; Rad54b; RDH54; RGD1306507
Gene Symbols RAD54B
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human RAD54B (aa 144-193).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25788
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.