Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RAD54L Antibody (Rad54 4E3/1), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB11013364MFV610
Description
RAD54L Monoclonal antibody specifically detects RAD54L in Human samples. It is validated for Flow CytometrySpecifications
RAD54L | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
Rad54 4E3/1 | |
Flow Cytometry | |
EC 3.6.1, hHR54EC 3.6.4.-, HR54, hRAD54HRAD54, RAD54 (S.cerevisiae)-like, RAD54 homolog, RAD54ADNA repair and recombination protein RAD54-like, RAD54-like (S. cerevisiae) | |
Full length human RAD54L protein | |
0.1 mL | |
DNA Repair, Homologous Recombination | |
8438 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction