Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RAD54L Monoclonal Antibody (60.1)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA123339
Description
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene belongs to the DEAD-like helicase superfamily, and shares similarity with Saccharomyces cerevisiae Rad54, a protein known to be involved in the homologous recombination and repair of DNA. This protein has been shown to play a role in homologous recombination related repair of DNA double-strand breaks. The binding of this protein to double-strand DNA induces a DNA topological change, which is thought to facilitate homologous DNA paring, and stimulate DNA recombination.
Specifications
RAD54L | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
Q92698 | |
RAD54L | |
Synthetic peptide - Combination of N & C peptide - Corresponding to a range between amino acids 1-300 and 450-741. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Western Blot | |
60.1 | |
Unconjugated | |
RAD54L | |
ATR2; DNA repair and recombination protein RAD54-like; h hRAD54; hHR54; HR54; hRAD54; MGC2094; mHR54; mRAD54; MRXHF1; Rad54; RAD54 homolog; RAD54 like (S. cerevisiae); RAD54A; RAD54L; RAD54-like (S. cerevisiae); RP5-875J14.1; SFM1; SHS; XH2; XNP; ZNF-HX | |
Mouse | |
Protein G | |
RUO | |
8438 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction