Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAD54L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565665
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565665 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565665 Supplier Invitrogen™ Supplier No. PA565665
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AKRKPEGRSCD DEDWQPGLVT PRKRKSSSET QIQECFLSPF RKPLSQLTNQ PPCLDSSQHE AFIRSILSKP FKVPIPNYQG PLGSRALGLK RAGVRRALHD PLEKDALVLY EPPPLSAHDQ LKLDKEKLPV HVVVDPILSK VL Highest antigen sequence identity to the following orthologs - mouse 92%, rat 88%.

The protein encoded by this gene belongs to the DEAD-like helicase superfamily, and shares similarity with Saccharomyces cerevisiae Rad54, a protein known to be involved in the homologous recombination and repair of DNA. This protein has been shown to play a role in homologous recombination related repair of DNA double-strand breaks. The binding of this protein to double-strand DNA induces a DNA topological change, which is thought to facilitate homologous DNA paring, and stimulate DNA recombination.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAD54L
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RAD54L
Gene Accession No. Q92698
Gene Alias ATR2; DNA repair and recombination protein RAD54-like; h hRAD54; hHR54; HR54; hRAD54; MGC2094; mHR54; mRAD54; MRXHF1; Rad54; RAD54 homolog; RAD54 like (S. cerevisiae); RAD54A; RAD54L; RAD54-like (S. cerevisiae); RP5-875J14.1; SFM1; SHS; XH2; XNP; ZNF-HX
Gene Symbols RAD54L
Host Species Rabbit
Immunogen Recombinant Human RAD54L. Recombinant protein control fragment (Product #RP-106339).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8438
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.