Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RAD54L2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA565253
Description
Immunogen sequence: EHGYPVSGGFA MPPVSLNHNL TTPFTSQAGE NSLFMGSTPS YYQLSNLLAD ARLVFPVTTD PLVPAGPVSS SSTATSVTAS NPSFMLNPSV PGIL Highest antigen sequence identity to the following orthologs - mouse 98%, rat 97%.
Adrenergic receptors (ARs) include four general types (a1, a2, b1 and b2) that are found in different target tissues and differ in their affinities and responses to various agonists and antagonists. The coupling of ARs to specific intracellular effectors is mediated through diverse heterotrimeric G proteins. ARs play a critical role in the development of prostate cancer, and transcriptional activity of AR is partly regulated by coregulatory proteins. RAD54L2 (RAD54-like 2), also known as ARIP4 (androgen receptor-interacting protein 4), HSPC325 or SRISNF2L, is a 1,467 amino acid nuclear protein belonging to the SNF2/RAD54 helicase family that consists of one helicase ATP-binding domain and a helicase C-terminal domain. RAD54L2 is a DNA helicase that regulates androgen receptor (AR)-dependent transactivation in a promoter-dependent manner. RAD54L2 is post-translationally sumoylated or phosphorylated upon DNA damage.
Specifications
RAD54L2 | |
Polyclonal | |
Unconjugated | |
RAD54L2 | |
Androgen receptor-interacting protein 4; AR interacting protein 4; ARIP4; Helicase ARIP4; HSPC325; KIAA0809; Rad54 like 2; RAD54L2; RAD54-like 2 (S. cerevisiae); RAD54-like protein 2; RGD1561889; SRISNF2L | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23132 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9Y4B4 | |
RAD54L2 | |
Recombinant Human RAD54L2. Recombinant protein control fragment (Product #RP-105971). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction