Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAD54Rabbit anti-Human, Polyclonal, Rockland™
SDP

Catalog No. 501991793
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-1793 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-199-1793 Supplier Rockland Immunochemicals Supplier No. 600401684
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. BLAST analysis indicates 100 % homology of the immunizing sequence with RAD54 from human. Cross reactivity with RAD54 protein homologues from other sources may not occur as sequence homology varies by at least one amino acid residue in this sequence.

The protein encoded by this gene belongs to the DEAD-like helicase superfamily, and shares similarity with Saccharomyces cerevisiae Rad54, a protein known to be involved in the homologous recombination and repair of DNA. This protein has been shown to play a role in homologous recombination related repair of DNA double-strand breaks. The binding of this protein to double-strand DNA induces a DNA topological change, which is thought to facilitate homologous DNA paring, and stimulate DNA recombination.

Specifications

Antigen RAD54
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene RAD54L
Gene Accession No. Q92698
Gene Alias ATR2; DNA repair and recombination protein RAD54-like; h hRAD54; hHR54; HR54; hRAD54; MGC2094; mHR54; mRAD54; MRXHF1; Rad54; RAD54 homolog; RAD54 like (S. cerevisiae); RAD54A; Rad54l; RAD54-like (S. cerevisiae); RP5-875J14.1; SFM1; SHS; XH2; XNP; ZNF-HX
Gene Symbols RAD54L
Host Species Rabbit
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8438
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.